missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Canine IGFBP-2 ELISA Kit

Canine IGFBP-2 quantitates canine IGFBP-2 in serum, plasma, supernatant.

Marca:  Invitrogen™ EC9RBX10

Detalles adicionales : Peso : 2.50000kg

 Ver más versiones de este producto

Código de producto. 17884040

  • 4540.00€ / paquete de 10

Fecha estimada de envío: 05-05-2023
para ver el stock
Añadir a la cesta

Descripción

Descripción

Canine IGFBP-2 quantitates canine IGFBP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant canine IGFBP-2.

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
Especificaciones

Especificaciones

AI255832; BP2; BRL-BP; IBP2; IBP-2; IGF binding protein 2; IGF-binding protein 2; IGFBP; IGFBP2; Igfbp-2; IGF-BP53; ILGFBPA; Insulin like growth factor binding protein; insulin like growth factor binding protein 2; insulin-like growth factor binding protein 2; insulin-like growth factor binding protein 2, 36 kDa; insulin-like growth factor-binding protein 2; mIGFBP-2; OTTHUMP00000207753
0.055 ng/mL
ELISA
Colorimetric Microplate Reader
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL
IGFBP2
4 hr 45 min
0.055 to 12.50 ng/mL
Biotin
Supernatant
IGFBP2
<10%
HRP
Research Use Only
2°C to 8°C
1 hr 20 min
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales