Learn More
Bismuth Subsalicylate, MedChemExpress
MedChemExpress Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.
Marca: Medchem Express HY-B0550
Descripción
Bismuth Subsalicylate is a potent and orally active antacid and anti-diarrheal agent. Bismuth Subsalicylate reduces inflammation/irritation of stomach and intestinal lining through inhibition of prostaglandin synthesis in vivo. Bismuth Subsalicylate is widely used for the research of diarrheal disorders, including indigestion, diarrhoea, nausea, et al.Especificaciones
Bismuth Subsalicylate | |
Blanco | |
C7H5BiO4 | |
Bismuth oxysalicylate Bismuth(III) salicylate basic | |
O=C1O[Bi](O)OC2=CC=CC=C12 | |
362.09 | |
Solid |
14882-18-9 | |
Research | |
10 g | |
DMSO : 1 mg/mL (2.76 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.28 mM; Need ultrasonic) | |
362.09 | |
Research | |
Neuroscience-Neuromodulation |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Research purposes only