Learn More
ACROBiosystems Biotinylated Human Transferrin R / CD71 Protein, His,Avitag™
MALS verified
Marca: ACROBiosystems TFR-H82E5-25ug

Descripción
CD antigen CD71 is also known as Transferrin receptor protein 1, TfR, sTfR, p90, TfR1, Trfr, which belongs to the peptidase M28 family and M28B subfamily. CD71 /TFR contains one PA (protease associated) domain. CD71 / TfR1 is required for iron delivery from transferrin to cells. CD71 is a potential new target in cases of human leukomia & lymphoma. CD71 /TFRC / TfR has been shown to interact with GABARAP and HFE.
This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 78.7 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_003225.2 | |
25 μg | |
Cys 89 - Phe 760 | |
CD71,TFRC,TFR,sTfR,TfR1,Trfr,T9,p90 | |
Precision Avi | |
Avi Tag and His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
78.7 kDa | |
Biotinylated Human Transferrin R, His,Avitag (TFR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1). | |
-20°C | |
Transferrin R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.