missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BCCIP Recombinant Protein
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
Especificaciones
Número de acceso | AAH09771 |
Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulación | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
ID de gen (Entrez) | 56647 |
Peso molecular | 60.17 |
Nombre | BCCIP (Human) Recombinant Protein (P01) |
Intervalo de pH | 8 |
Método de preparación | In vitro wheat germ expression system |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ BCCIP Recombinant Protein >
Spot an opportunity for improvement?Share a Content Correction