Learn More
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67057-1-IG-20UL
 Ver más versiones de este producto
Código de producto. 16889455
  • 145.00€ / 20 microlitros
Descripción
The ATX3, ATXN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATX3, ATXN3. This antibody recognizes human, mouse, pig antigen. The ATX3, ATXN3 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
ATX3 ATXN3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig, Mouse
Western Blot, ELISA
1F7E10
Unconjugated
20 μL
4287
IgG2a
Certificados
Promociones
Promociones
Título del producto
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech > 20 μL