Learn More
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67057-1-IG-150UL
 Ver más versiones de este producto
Código de producto. 16879455
  • 410.00€ / 150 microlitros
Descripción
The ATX3, ATXN3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ATX3, ATXN3. This antibody recognizes human, mouse, pig antigen. The ATX3, ATXN3 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
Especificaciones
ATX3 ATXN3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig, Mouse
Western Blot, ELISA
1F7E10
Unconjugated
150 μL
4287
IgG2a
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
ATX3 ATXN3 Mouse anti-Human, Unconjugated, Clone: 1F7E10, Proteintech > 150 μL