Learn More
ARH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Especificaciones
Antígeno ARH3
Concentración 0.18 mg/mL
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16816164
Ver documentos
Proteintech
16504-1-AP-20UL
20 μL
145.00€
20 microlitros
16806164
Ver documentos
Proteintech
16504-1-AP-150UL
150 μL
410.00€
150 microlitros
Descripción
Descripción
ADPRHL2 is a member of the ADP-ribosylglycohydrolase family. The enzyme catalyzes the removal of ADP-ribose from ADP-ribosylated proteins. This enzyme localizes to the mitochondria, in addition to the nucleus and cytoplasm.
Especificaciones
Especificaciones
ARH3
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q8CG72, Q9NX46
100206, 362600, 54936
ARH3 Fusion Protein Ag9683
Primary
-20°C
ADPRHL2
0.18 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ADP ribosylhydrolase 3, ADP ribosylhydrolase like 2, ADPRHL2, ARH3
Adprhl2
IgG
Antigen Affinity Chromatography
Antibody
Documentos
Product Certifications
Título del producto
ARH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech