Learn More
Abnova™ APBB1 (Human) Recombinant Protein (P01)
Human APBB1 full-length ORF with GST-tag at N-terminal
Marca: Abnova™ H00000322-P01.10ug
Descripción
Human APBB1 full-length ORF ( NP_663722.1, 1 a.a. - 708 a.a.) recombinant protein with GST-tag at N-terminal
- Theoretical MW: 103.4kDa
- Purification Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Best use within three months from the date of receipt of this protein
Enzyme Linked Immunosorbent Assay, Western Blotting, Antibody Production, Protein Array
Especificaciones
NP_663722.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
103.4 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APBB1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
322 | |
APBB1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
FE65/MGC:9072/RIR | |
APBB1 | |
Wheat Germ (in vitro) | |
GST |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.