Learn More
ADRP/Perilipin 2 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech CL488-15294-20UL
 Ver más versiones de este producto
Código de producto. 17791259
  • 145.00€ / 20 microlitros
Descripción
ADRP/Perilipin 2 Polyclonal antibody specifically detects ADRP/Perilipin 2 in Human, Mouse, Rat samples. It is validated for IF
Especificaciones
Especificaciones
ADRP/Perilipin 2
Polyclonal
CoraLite 488
BC029886
Antigen affinity purification
Primary
Human, Mouse, Rat
Antibody
IgG
Immunofluorescence
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Rabbit
20 μL
123
Store at -20°C
Liquid
Certificados
Promociones
Promociones
Título del producto
ADRP/Perilipin 2 Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech > 20 μL; CoraLite 488