missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD7 Monoclonal Antibody (08), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543044
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
Specificity: Human ACBD7
Binds medium- and long-chain acyl-CoA esters.
Specifications
ACBD7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8N6N7 | |
ACBD7 | |
Recombinant Human ACBD7 Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
08 | |
Unconjugated | |
ACBD7 | |
9230116B18Rik; ACBD7; acyl-CoA binding domain containing 7; acyl-CoA-binding domain-containing protein 7; acyl-Coenzyme A binding domain containing 7; bA455B2.2; RGD1564164 | |
Mouse | |
Protein A | |
RUO | |
414149 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACBD7 Monoclonal Antibody (08), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction