missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ A20/TNFAIP3 Recombinant Protein Antigen
Recombinant Protein
Marca: Novus Biologicals™ NBP2-68857PEP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción

Especificaciones
Antibody Competition | |
7128 | |
100 μL | |
A20TNFA1P2, EC 3.4.19.12, EC 6.3.2.-, MGC104522, MGC138687, OTU domain-containing protein 7 C, OTUD7CMGC138688, Putative DNA-binding protein A20, TNF alpha-induced protein 3, tumor necrosis factor alpha-induced protein 3, tumor necrosis factor inducible protein A20, tumor necrosis factor, alpha-induced protein 3, Zinc finger protein A20 | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-68857. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL | |
>80% by SDS-PAGE and Coomassie blue staining |
PBS and 1 M Urea, pH 7.4. | |
MolecularWeight-theroretical: 28 kDa | |
Store at -20°C. Avoid freeze/thaw cycles. | |
TNFAIP3 | |
Cancer, Inflammation, Innate Immunity, Necroptosis, Neurodegeneration, Signal Transduction | |
A20/TNFAIP3 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ A20/TNFAIP3 Recombinant Protein Antigen > 100 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido