Learn More
R&D Systems™ Recombinant Human R-spondin 1, Animal-Free Protein
Marca: R&D Systems™ Qk006-0050
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
No significant difference between EC50 of reference and test lots
Especificaciones
Q2MKA7.1 | |
Lyophilized from acetonitrile/TFA | |
M.W. (Observed): Monomeric R-spondin 1 protein only, M.W. (Theroretical): 13 kDa | |
E. coli-derived human R-Spondin 1 protein | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Unconjugated | |
R-Spondin 1 |
Bioactivity | |
284654 | |
50 μg | |
Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles. | |
CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPONDIN, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired. |
Título del producto
R&D Systems™ Recombinant Human R-spondin 1, Animal-Free Protein > 50 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido