missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAP70 Antibody (ZAP70/528), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-11537CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ZAP70 Monoclonal antibody specifically detects ZAP70 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Especificaciones
ZAP70 | |
Monoclonal | |
CoraFluor 1 | |
70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD | |
Recombinant full-length human ZAP70 protein (Uniprot: P43403) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
7535 | |
Store at 4°C in the dark. Do not freeze. | |
IgG2a κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ZAP70/528 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
ZAP70 Antibody (ZAP70/528), CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido