Learn More
YAP1 Mouse anti-Human, CoraLite 488, Clone: 3A7A9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66900-100UL
Descripción
The YAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YAP1. This antibody recognizes human, mouse, rat antigen. The YAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
YAP1 | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Immunofluorescence | |
3A7A9 | |
CoraLite 488 | |
100 μL | |
10413 | |
IgG1 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.