Learn More
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper Conformity Certified is thick-walled, clearly marked and labeled, and has a solid finish
Marca: Karl Hecht™ 42096084
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Duran™ borosilicate glass 3.3.
- Standard type.
- Standard ground and interchangeable NS stopper of polyethylene.
- Fused blue graduation.
- Thick-walled, solid finish.
- Clearly legible figures and marks.
- Conformity certified.
- Individual test- or lot-control certificate available from manufacturer with surcharge.
Especificaciones
DIN EN ISO 1042 | |
1000 mL | |
Standard | |
0.6 mL | |
Tapón NS |
Polietileno | |
Borosilicate Glass (3.3) | |
Volumetric Flask | |
Sí | |
NS 29/32 |
Título del producto
Karl Hecht™ Volumetric Flask with PP Stopper, Conformity Certified > Capacity: 1000mL, Closure Size: NS 29/32, Neck Style: Standard, Tolerance: 0.6mL
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido