missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VNN2 Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA543420
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human VNN2/Vanin-2
This gene product is a member of the Vanin family of proteins that share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Especificaciones
VNN2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O95498 | |
VNN2 | |
Recombinant Human VNN2/Vanin-2 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
018 | |
Unconjugated | |
VNN2 | |
FOAP-4; Glycosylphosphatidyl inositol-anchored protein GPI-80; GPI-80; LOW QUALITY PROTEIN: vascular non-inflammatory molecule 2; pantetheinase; Protein FOAP-4; vanin 2; Vanin-2; Vannin 2; vascular non-inflammatory molecule 2; VNN2 | |
Rabbit | |
Protein A | |
RUO | |
8875 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
VNN2 Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido