missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Mouse anti-Human, BV605, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 749653
539.91 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
VISTA Monoclonal antibody specifically detects VISTA in Human samples. It is validated for Flow Cytometry
Especificaciones
VISTA | |
Monoclonal | |
Brilliant Violet 605 | |
VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1 | |
Human VISTA Transfected Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
MIH65.rMAb (also known as MIH65) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV605 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
VISTA Mouse anti-Human, BV605, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™ > 50 μg; BV605
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido