Learn More
TPM4 Mouse anti-Human, Unconjugated, Clone: 2E12D12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67244-1-IG-150UL
Descripción
The TPM4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPM4. This antibody recognizes human, mouse, rat, pig antigen. The TPM4 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
TPM4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
2E12D12 | |
Unconjugated | |
150 μL | |
7171 | |
IgG1 |