missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGIT Recombinant Mouse Monoclonal Antibody (1B4)
Mouse Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA548198
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: This antibody is specific for murine TIGIT.
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Especificaciones
TIGIT | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P86176 | |
TIGIT | |
This antibody was raised by immunising TIGIT-/- mice with recombinant murine TIGIT tetramers. | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 λ |
ELISA, Flow Cytometry, Inhibition Assays | |
1B4 | |
Unconjugated | |
TIGIT | |
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
Mouse | |
Protein A | |
RUO | |
100043314 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ TIGIT Recombinant Mouse Monoclonal Antibody (1B4) > 200 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido