missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie2 (CD202b) Mouse anti-Human, Mouse, Alexa Fluor 647, Clone: 33 (also known as MoAb33; Ab33), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566716
315.31 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Tie2 (CD202b) Monoclonal antibody specifically detects Tie2 (CD202b) in Human, Mouse samples. It is validated for Flow Cytometry
Especificaciones
Tie2 (CD202b) | |
Monoclonal | |
Alexa Fluor 647 | |
CD202B; TIE-2; TIE2; GLC3E; TEK; TEK tyrosine kinase, endothelial; VMCM; VMCM1 | |
Human TIE2 extracellular domain Recombinant Protein | |
RUO | |
Human, Mouse | |
IgG1 κ |
Flow Cytometry | |
33 (also known as MoAb33; Ab33) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Tie2 (CD202b) Mouse anti-Human, Mouse, Alexa Fluor 647, Clone: 33 (also known as MoAb33; Ab33), BD Biosciences™ > 0.1 mg; Alexa Fluor 647
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido