missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 3 Antibody (JOVI.3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-50356-0.025ml
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TCR V beta 3 Monoclonal antibody specifically detects TCR V beta 3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF
Especificaciones
TCR V beta 3 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Functional, CyTOF-ready | |
P09693 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional Assay, CyTOF | |
JOVI.3 | |
Unconjugated | |
PBS | |
T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
Thymus, spleen and mesenteric lymph nodes isolated from a mouse transgenic for human Vb3 TcR. | |
0.025 mL | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
6957 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
TCR V beta 3 Antibody (JOVI.3) - BSA Free, Novus Biologicals™ > 0.025 mL, Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido