missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAOK2 (TAO1) Recombinant Human Protein
TAOK2 (TAO1) is a serine/threonine kinase.
Marca: Invitrogen™ PV4068
Detalles adicionales : Peso : 0.00900kg
Descripción
TAOK2 (TAO1) may be an important regulator of the stress-responsive p38 MAP kinase pathway.
Especificaciones
NP_004774 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
TAOK2 (TAO1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
63.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |