missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK3 (MST2) Recombinant Human Protein
STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways.
Marca: Invitrogen™ PR8881A
Detalles adicionales : Peso : 0.01000kg
Descripción
STK3 (MST2) is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand.
Especificaciones
NP_006272.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
STK3 (MST2) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
85.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |