Learn More
Invitrogen™ Staphylococcus aureus Monoclonal Antibody (Staph12-569.3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA110710
Descripción
MA1-10710 detects Staphylococcus aureus from Staph. aureus and Staph. epidermidis samples. MA1-10710 has been successfully used in ELISA applications. The MA1-10710 immunogen is UV-inactivated Staphylococcus aureus cells.
Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.
Especificaciones
Staphylococcus aureus | |
Monoclonal | |
2.8 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Purified | |
RUO | |
Bacteria | |
Antibody | |
IgG3 |
ELISA | |
Staph12-569.3 | |
Unconjugated | |
MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant | |
UV-inactivated Staphylococcus aureus cells. | |
500 μg | |
Primary | |
4°C, do not freeze | |
Liquid |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
79607611 | Certificado de análisis | 23/07/2025 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.