Learn More
Solución de tiocianato de sodio, 0,1mol/L, Fisher Chemical™
Shop All Fisher Chemical Products
missing translation for 'orderingAttributeHoverText'
Cantidad:
1000 mL
Envase:
Frasco
missing translation for 'unitSize'
1000 ml
Specifications
Specifications
Nombre del producto químico o material | Sodium Thiocyanate |
CAS | 7732-18-5 |
Cantidad | 1000 mL |
Sinónimo | dihydrogen oxide, dihydrogen monoxide |
Concentration or Composition (by Analyte or Components) | 0.1M (0.1N) |
Envase | Frasco |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Solución de tiocianato de sodio, 0,1mol/L, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction