Learn More
SIL1 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marca: Origene Technologies TA500996

Descripción
This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.Especificaciones
SIL1 | |
Monoclonal | |
0.59 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H173 | |
SIL1 | |
Full length human recombit protein of human SIL1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1G7 | |
Unconjugated | |
SIL1 | |
BAP, MSS, ULG5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64374 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.