Learn More
Siglec-7/CD328 Mouse anti-Human, Clone: 194212, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Marca: R&D Systems MAB1138-SP

Descripción
Siglec-7/CD328 Monoclonal specifically detects Siglec-7/CD328 in Human samples. It is validated for Western Blot.Especificaciones
Siglec-7/CD328 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9Y286 | |
SIGLEC7 | |
Mouse myeloma cell line NS0-derived recombinant human Siglec-7/CD328 Gln19-Gly357 (predicted) Accession # Q9Y286 | |
25 μg | |
Primary | |
Detects human Siglec-7/CD328 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Siglec-2, -3, -5, or -9 is observed. | |
Human | |
Purified |
Western Blot | |
194212 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Adhesion inhibitory receptor molecule 1, adhesion inhibitory receptor molecule 1, siglec-7, AIRM1, AIRM-1, AIRM1QA79 membrane protein, CD328, CD328 antigen, CDw328, D-siglec, p75, p75/AIRM1, QA79, sialic acid binding Ig-like lectin 7, sialic acid binding immunoglobulin-like lectin 7, sialic acid-binding Ig-like lectin 7, Siglec7, Siglec-7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27036 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.