Learn More
Sestrin 2 Mouse anti-Human, CoraLite 594, Clone: 1A1A4, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL594-66297-100UL
Código de producto. 17224806
  • 450.00€ / 100 microlitros
Explore más ofertas especiales
Descripción
Descripción
Sestrins, including sestrin-1 (PA26), sestrin-2 (Hi95), and sestrin-3, are 48 to 65 kDa cystein sulfinyl reductases and they modulate peroxide signaling and antioxidant defense. These proteins selectively reduce or repair hyperoxidized forms of typical 2-Cys peroxiredoxins within eukaryotes. Expression of these proteins is regulated by p53, a tumor suppressor protein. Sestrin 2 is implicated in the regulation of cell survival, in cellular responses to different stress conditions, and also acts as a positive regulator of autophagy. Sestrin 2 is approximately a 57.6kDa protein (480 amino acids).
Especificaciones
Especificaciones
Sestrin 2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG1
Immunofluorescence
1A1A4
CoraLite 594
100 μL
83667
Liquid
Documentos
One moment while we fetch your results.
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
Sestrin 2 Mouse anti-Human, CoraLite 594, Clone: 1A1A4, Proteintech > 100 μL