Learn More
Sestrin 2 Mouse anti-Human, CoraLite 594, Clone: 1A1A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL594-66297-100UL
Descripción
Sestrins, including sestrin-1 (PA26), sestrin-2 (Hi95), and sestrin-3, are 48 to 65 kDa cystein sulfinyl reductases and they modulate peroxide signaling and antioxidant defense. These proteins selectively reduce or repair hyperoxidized forms of typical 2-Cys peroxiredoxins within eukaryotes. Expression of these proteins is regulated by p53, a tumor suppressor protein. Sestrin 2 is implicated in the regulation of cell survival, in cellular responses to different stress conditions, and also acts as a positive regulator of autophagy. Sestrin 2 is approximately a 57.6kDa protein (480 amino acids).
Especificaciones
Sestrin 2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human | |
IgG1 |
Immunofluorescence | |
1A1A4 | |
CoraLite 594 | |
100 μL | |
83667 | |