Learn More
VACUUBRAND™ Filtros de cono seguro Biohit™
Evita que aerosoles o líquidos contaminen muestras y pipetas. Los filtros Safe-Cone Sartorius™ Biohit™ son filtros de polietileno únicos y reemplazables que aumentan la eficiencia del pipeteo y la integridad de las muestras al mismo tiempo. Disponibles en versiones estándar y Plus para diferentes aplicaciones de laboratorio.
Marca:  VACUUBRAND™ 20721005
Código de producto. 11939941
  • 956.80€ / cada uno
Solo quedan null
Fecha estimada de envoi: 30-07-2025
Descripción
Protection for pipets and samples
  • Acts as a final barrier to the internal components of the pipet
  • Protects the pipet and the sample from contamination
  • Prolongs the pipet’s lifetime and reduces maintenance intervals
  • Cost-effective, replaceable filters constructed of polyethylene
Available in two formats:
  • Standard Safe-Cone Filters: For general applications and can be used for the same type of work as the Plus Filter, but need to be changed more frequently
  • Plus Safe-Cone Filters: Self-sealing filters meant for more demanding applications, such as radioactive work, cell culture, bacterial and virological work, and molecular biology
Change the filter as needed
  • The frequency of changing the filter depends on the application and the sample
  • Should be changed daily (or after 50 to 250 pipettings) or immediately in case of over-aspiration
  • To ensure safety of the user, forceps should be used to change the filter
  • In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter
Documentos
One moment while we fetch your results.
Certificados
Promociones
Promociones
Título del producto
VACUUBRAND™ Filtros de cono seguro Biohit™ > Type: Standard; Dia.: 6.73mm