missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Sartopore™ 2 MidiCaps Polypropylene Housing Polyethersulfone Membrane Filter Capsules
Marca: Sartorius™ 5445307H9--SS--A
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción

Especificaciones
Autoclavable | |
Double Layer PES | |
Sterile | |
For filtration of biologicals, pharmaceuticals, cell culture media and components and buffers | |
0.5 in. Tri-Clamp | |
0.5 in. Tri-Clamp | |
Filter Capsule | |
77 mm |
ISO 9001, USP Class VI | |
0.2 μm | |
0.05 m2 | |
Polypropylene | |
1Each | |
1 Each | |
197 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius™ Sartopore™ 2 MidiCaps Polypropylene Housing Polyethersulfone Membrane Filter Capsules > Length: 197mm; Filtration Rate: 0.05m&superscript_2;; Outlet Style: 0.5 in. Tri-Clamp; Pore Size: 0.2μm
Spot an opportunity for improvement?Share a Content Correction