missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike RBD Antibody (B001N) - (RBD) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals NBP3-05728
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SARS-CoV-2 Spike RBD Monoclonal antibody specifically detects SARS-CoV-2 Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Neutralization
Especificaciones
SARS-CoV-2 Spike RBD | |
Monoclonal | |
Unconjugated | |
PBS, pH7.2 | |
Human | |
Protein A purified | |
RUO | |
Primary | |
SARS-CoV-2 | |
Purified |
Western Blot, ELISA, Neutralization | |
B001N | |
Western Blot 1:1000-2000, ELISA 1:320000, Block/Neutralize | |
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein | |
SARS-CoV-2 Spike RBD recombinant protein | |
50 μg | |
Infections (Virus Bacteria and Parasites) | |
43740568 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
SARS-CoV-2 Spike RBD Antibody (B001N) - (RBD) - Azide and BSA Free, Novus Biologicals™ > 50 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido