Learn More
ACROBiosystems SARS-CoV-2 S1 protein (HV69-70del, Y144del, N501Y, A570D, D614G, P681H), Fc Tag
Marca:  ACROBiosystems S1D-C5254-1mg
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 17216440
  • 3411.22€ / 1 miligramo
Fecha estimada de envío: 09-05-2024
para ver el stock
Descripción
Descripción
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.1 kDa.
Especificaciones
Especificaciones
101.1 kDa
SARS-CoV-2 S1 protein, Fc Tag (S1D-C5254) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).
-20°C
S1 protein
SARS-CoV-2
HEK293
Viral Proteins
1 mg
Val 16 - Arg 685
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
Fc Tag
Powder