Learn More
Checkers Safety Rider™ Mini Humps
Safety Rider™ Mini Humps are a unique modular traffic calming device designed to slow traffic while maintaining a continuous traffic flow.
138.00€
Especificaciones
Material | Caucho reciclado |
---|---|
Longitud (imperial) | 35 in |
Longitud (métrico) | 90 cm |
Anchura (métrico) | 50 cm |
Código de producto | Marca | Color | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Color | Precio | Cantidad y disponibilidad | |||||
17702328
|
Checkers
GNRS3500YB |
Negro, Amarillo |
138.00€
cada uno |
Fecha estimada de envío: 14-05-2024
Inicie sesión para ver el stock |
|||||
17792318
|
Checkers
GNRS3500WB |
Negro, Blanco |
Este artículo actualmente no está disponible o ha sido discontinuado. Ver la página del producto para posibles alternativas |
||||||
Descripción
- Safety Rider™ Mini Humps are a unique modular traffic calming device designed to slow traffic while maintaining a continuous traffic flow
- The mini hump is constructed of interlocking units with a tongue and groove system
- This allows the humps to be linked together to cover the width of any roadway
- End caps are available to provide a smooth finish to the ends of the hump
- The Safety Rider™ is constructed from 100% recycled tires
- Unlike asphalt humps, our mini humps are preformed for a uniform and consistent profile and a smooth, safe ride
- The mini humps are simple to install and do not require road closures
- Plastic shields are inserted into pre-drilled holes and the units are bolted down to keep from shifting
- The Safety Rider™ can be easily removed for storage or relocation
- Mini speed humps are ideal for slowing traffic in school parking lots and crossings, playground areas, hospitals, parking facilities, and anywhere a slow traffic flow is required
- Heavy load bearing truck traffic is ideally facilitated by the Safety Rider™ mini hump
- Moulded highly reflective tape provides optimal visibility even on a dark night, alerting drivers well in advance
- Safety Rider™ mini humps are covered by a 3-year warranty; 1 year on the tape (under normal use)
Especificaciones
Caucho reciclado | |
35 in | |
90 cm | |
50 cm |