Learn More
Invitrogen™ RUNX3 Monoclonal Antibody (2B3)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA517169
Descripción
MA5-17169 targets RUNX3 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17169 immunogen is purified recombinant fragment of human RUNX3 (amino acids:186-252) expressed in E. Coli. MA5-17169 detects RUNX3 which has a predicted molecular weight of approximately 44.4kDa.
This gene encodes a member of the runt domain-containing family of transcription factors. A heterodimer of this protein and a beta subunit forms a complex that binds to the core DNA sequence 5'-PYGPYGGT-3' found in a number of enhancers and promoters, and can either activate or suppress transcription. It also interacts with other transcription factors. It functions as a tumor suppressor, and the gene is frequently deleted or transcriptionally silenced in cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
RUNX3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q13761, Q64131 | |
RUNX3 | |
Purified recombinant fragment of human RUNX3 (amino acids:186-252) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2B3 | |
Unconjugated | |
RUNX3 | |
Acute myeloid leukemia 2 protein; acute myeloid leukemia gene 2; AML2; CBFA3; CBF-alpha 3; CBF-alpha-3; core binding factor alpha 3; core-binding factor subunit alpha-3; core-binding factor, runt domain, alpha subunit 3; oncogene AML-2; PEA2 alpha C; PEA2-alpha C; PEBP2 alpha C; PEBP2A3; PEBP2aC; PEBP2-alpha C; polyomavirus enhancer-binding protein 2 alpha C subunit; runt domain, alpha subunit 3; Runt related transcription factor 3; runt-related transcription factor 3; RUNX3; SL3/AKV core-binding factor alpha C subunit; SL3-3 enhancer factor 1 alpha C subunit; transcription factor AML2; transcription factor AML2/CBFA3 | |
Mouse | |
Protein G | |
RUO | |
12399, 864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
2012232668 | Certificado de análisis | 21/07/2025 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.