Learn More
RIPK1/RIP1 Antibody (OTI2D6), Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Bio-Techne NBP2-45591
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
RIPK1/RIP1 Monoclonal specifically detects RIPK1/RIP1 in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
RIPK1/RIP1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
Mouse | |
75.8 kDa | |
0.1 mL | |
Apoptosis, Necroptosis | |
8737 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2D6 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RIPK1 | |
Human recombinant protein fragment corresponding to amino acids 133-422 of human RIPK1(NP_003795) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Canine, Monkey | |
Purified |
Título del producto
RIPK1/RIP1 Antibody (OTI2D6), Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido