Learn More
ACROBiosystems Rhesus macaque CD155 / PVR Protein, His Tag
399.17€ - 2896.79€
Especificaciones
Fuente | Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession # Q0MSE6-1). |
---|---|
Nombre común | CD155 |
Sistema de expresión | HEK293 |
Formato | Powder |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16477309
|
ACROBiosystems
CD5-R52H5-1MG |
1 mg |
2896.79€
1 miligramo |
Fecha estimada de envío: 14-09-2022 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. |
16467309
|
ACROBiosystems
CD5-R52H5-100UG |
100 µg |
399.17€
100 microgramos |
Fecha estimada de envío: 14-09-2022 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. |
Descripción
CD155 (cluster of differentiation 155) also known as the poliovirus receptor is a protein that is encoded by the PVR gene. CD155 is a Type I transmembrane glycoprotein in the immunoglobulin superfamily. Commonly known as Poliovirus Receptor (PVR) due to its involvement in the cellular poliovirus infection in primates, CD155's normal cellular function is in the establishment of intercellular adherens junctions between epithelial cells. The role of CD155 in the immune system is unclear, though it may be involved in intestinal humoral immune responses. Subsequent data has also suggested that CD155 may also be used to positively select MHC-independent T cells in the thymus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.3 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession # Q0MSE6-1). | |
HEK293 | |
Unconjugated | |
-20°C |
CD155 | |
Powder | |
Immune Checkpoint Proteins |