Learn More
ACROBiosystems Rhesus macaque CD155 / PVR Protein, His Tag
Marca: ACROBiosystems CD5-R52H5-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
CD155 (cluster of differentiation 155) also known as the poliovirus receptor is a protein that is encoded by the PVR gene. CD155 is a Type I transmembrane glycoprotein in the immunoglobulin superfamily. Commonly known as Poliovirus Receptor (PVR) due to its involvement in the cellular poliovirus infection in primates, CD155's normal cellular function is in the establishment of intercellular adherens junctions between epithelial cells. The role of CD155 in the immune system is unclear, though it may be involved in intestinal humoral immune responses. Subsequent data has also suggested that CD155 may also be used to positively select MHC-independent T cells in the thymus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.3 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001036851.1 | |
1 mg | |
Asp 28 - Asn 343 | |
PVR,FLJ25946,PVS,CD155,TAGE4,HVED,NECL5 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
36.3 kDa | |
Rhesus macaque CD155, His Tag (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession # Q0MSE6-1). | |
-20°C | |
CD155 | |
Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |