missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Virus Norovirus Group-2 Protein
A cDNA sequence encoding the Norovirus Group-2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12867-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Virus Norovirus Group-2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKMASNDATP SNDGAAGLVP EINNEAMALD PVAGAAIAAP LTGQQNIIDP WIMNNFVQAP GGEFTVSPRN SPGEVLLNLE LGPEINPYLA HLARMYNGYA GGFEVQVVLA GNAFTAGKII FAAIPPNFPI ENLSAAQITM CPHVIVDVRQ LEPVNLPMPD VRNNFFHYNQ GSDSKLRLVA MLYTPLRANN SGDDVFTVSC RVLTRPSPDF SFNFLVPPTV ESKTKPFTLP ILTISEMSNS RFPVPIDSLH TSPTENIVVQ CQNGRVTLDG ELMGTTQLLP SQICAFRGVL TRSTSRASDQ ADTATPRLFN YYWHIQLDNL NGTPYDPAED IPGPLGTPDF RGKVFGVASQ RNPDSTTRAH EAKVDTTAGR FTPKLGSLEI STESGDFDQN QPTRFTPVGI GVDHEADFQQ WSLPDYSGQF THNMNLAPAV APNFPGEQLL FFRSQLPSSG GRSNGILDCR VPQEWVQHFY QESAPAQTQV ALVRYVNPDT GRVLFEAKLH KLGFMTIAKN GDSPITVPPN GYFRFESWVN PFYTLAPMGT GNGRRRIQ | |
Protein is >85% pure as determined by 12% PAGE (coomassie staining). |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Virus | |
His | |
Phosphate buffer, 1M urea and 50mM arginine. |