missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae HYP2 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) HYP2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5977-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_010880.3 | |
S. cerevisiae HYP2 Protein | |
Research Use Only | |
HYP2 | |
Recombinant Protein | |
E. coli | |
SDEEHTFETA DAGSSATYPM QCSALRKNGF VVIKSRPCKI VDMSTSKTGK HGHAKVHLVA IDIFTGKKLE DLSPSTHNME VPVVKRNEYQ LLDIDDGFLS LMNMDGDTKD DVKAPEGELG DSLQTAFDEG KDLMVTIISA MGEEAAISFK EAARTD | |
Greater than 90% as determined by SDS-PAGE. |
856677 | |
50 μg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |