missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Enterotoxin type B Protein
A DNA sequence encoding the Staphylococcus aureus Enterotoxin type B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7344-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_000278085.1 | |
S. aureus Enterotoxin type B Protein | |
Research Use Only | |
entB | |
Recombinant Protein | |
Yeast | |
ESQPDPKPDE LHKSSKFTGL MENMKVLYDD NHVSAINVKS IDQFLYFDLI YSIKDTKLGN YDNVRVEFKN KDLADKYKDK YVDVFGANYY YQCYFSKKTN DINSHQTDKR KTCMYGGVTE HNGNQLDKYR SITVRVFEDG KNLLSFDVQT NKKKVTAQEL DYLTRHYLVK NKKLYEFNNS PYETGYIKFI ENENSFWYDM MPAPGDKFDQ SKYLMMYNDN KMVDSKDVKI EVYLTTKKK | |
Greater than 80% as determined by SDS-PAGE. |
31213656 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
S. aureus | |
His | |
Tris-based buffer, 50% glycerol |