missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat VEGFA-164 Protein
Recombinant Protein
Marca: enQuireBio™ QP5217-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001274037.1 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat VEGFA-164 bioactivity tested in HUVEC assay. Rat VEGFA-164 induces HUVEC proliferation (typical ED50 is ? 10 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
VEGFA-164 Protein | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
E. coli | |
MAPTTEGEQK AHEVVKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNVTMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR | |
Greater than 95% as determined by SDS-PAGE. |