missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Type-2 angiotensin II receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Type-2 angiotensin II receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5626-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036626.1 | |
Type-2 angiotensin II receptor Protein | |
Research Use Only | |
Agtr2 | |
Recombinant Protein | |
E. coli | |
MKDNFSFAAT SRNITSSLPF DNLNATGTNE SAFNCSHKPA DKHLE | |
Greater than 90% as determined by SDS-PAGE. |
24182 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |