missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Prolactin / PRL Protein
A DNA sequence encoding the Rat Prolactin / PRL, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10154-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036761.1 | |
Prolactin / PRL Protein | |
Research Use Only | |
PRL | |
The ED50 as determined by a cell proliferation assay using rat Nb2-11 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
24683 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LPVCSGGDCQ TPLPELFDRV VMLSHYIHTL YTDMFIEFDK QYVQDREFIA KAINDCPTSS LATPEDKEQA QKVPPEVLLN LILSLVHSWN DPLFQLITGL GGIHEAPDAI ISRAKEIEEQ NKRLLEGIEK IISQAYPEAK GNEIYLVWSQ LPSLQGVDEE SKDLAFYNNI RCLRRDSHKV DNYLKFLRCQ IVHKNNC | |
Greater than 98% as determined by SDS-PAGE and HPLC. |