missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Neuroendocrine convertase 2 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Neuroendocrine convertase 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6473-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036878.1 | |
Neuroendocrine convertase 2 Protein | |
Research Use Only | |
Pcsk2 | |
Recombinant Protein | |
E. coli | |
GYRDINEIDI NMNDPLFTKQ WYLFNTGQAD GTPGLDLNVA EAWELGYTGK GVTIGIMDDG IDYLHPDLAY NYNSDASYDF SSNDPYPYPR YTDDWFNSHG TRCAGEVSAA ASNNICGVGV AYNSKVAGIR MLDQPFMTDI IEASSISHMP QLIDIYSASW GPTDNGKTVD GPRELTLQAM ADGVNKGRGG KGSIYVWASG DGGSYDDCNC DGYASSMWTI SINSAINDGR TALYDESCSS TLASTFSNGR KRNPEAGVAT TDLYGNCTLR HSGTSAAAPE AAGVFALALE ANVDLTWRDM QHLTVLTSKR NQLHDEVHQW RRNGVGLEFN HLFGYGVLDA GAMVKMAKDW KTVPERFHCV GGSVQNPEKI PPTGKLVLTL QTNACEGKEN FVRYLEHVQA VITVNATRRG DLNINMTSPM GTKSILLSRR PRDDDSKVGF DKWPFMTTHT WGEDARGTWT LELGFVGSAP QKGLLKEWTL MLHGTQSAPY IDQVVRDYQS KLAMSKKQEL EEELDEAVER SLQSILRKN | |
Greater than 90% as determined by SDS-PAGE. |
25121 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |