missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat LIF Protein
A cDNA sequence encoding the LIF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10771-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
60584 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity of rat LIF is determined by the ability to induce differentiation of M1 myeloid leukemic cells. The minimum detectable concentration of rat LIF in this assay is 0.5ng/mL. | |
Rat | |
Untagged | |
LIF Rat was lyophilized from 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |
LIF Protein | |
Research Use Only | |
Lif | |
Recombinant Protein | |
E. coli | |
SPLPITPVNA TCAIRHPCHG NLMNQIKSQL AQLNGSANAL FISYYTAQGE PFPNNVDKLC APNMTDFPPF HANGTEKTKL VELYRMVTYL GASLTNITWD QKNLNPTAVS LQIKLNATTD VMRGLLSSVL CRLCNKYHVG HVDVPCVPDN SSKEAFQRKK LGCQLLGTYK QVISVLAQAF | |
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |