missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL-17AF Protein
Recombinant Protein
Marca: enQuireBio™ QP5206-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL-17AF Protein | |
Research Use Only | |
IL17 | |
Recombinant Protein | |
E. coli | |
IL-17A:MAV LIPQSSVCPN AEANNFLQNV KVNLKVLNSL SSKASSRRPS DYLNRSTSPW TLSRNEDPDR YPSVIWEAQC RHQRCVNAEG KLDHHMNSVL IQQEILVLKR EPEKCPFTFR VEKMLVGVGC TCVSSIVRHA S IL-17F:MA RRNPKVGLSA LQKAGNCPPL EDNSVRVDIR IFNQNQGISV PRDFQNRSSS PWDYNITRDP DRFPSEIAEA QCRHSGCINA QGQEDGSMNS VPIQQEILVL RREPQGCSNS FRLEKMLIKV GCTCVTPIVH HAA | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL-17AF induces IL-6 production in 3T3 cells as detected via ELISA (each lot is tested and typical ED50 is less than 20 ng/ml). | |
Rat | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |