missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Complement C1q subcomponent subunit A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Complement C1q subcomponent subunit A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5736-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001008515.1 | |
Complement C1q subcomponent subunit A Protein | |
Research Use Only | |
C1QA | |
Recombinant Protein | |
E. coli | |
EDVCRAPNGK DGVAGIPGRP GRPGLKGERG EPGAAGIRTG IRGLKGDMGE SGPPGKPGNV GFPGPTGPLG NSGPQGLKGV KGNPGNIRDQ PRPAFSAIRQ NPPTYGNVVV FDKVLTNQEN PYQNRTGHFI CAVPGFYYFT FQVISKWDLC LSIVSSSRGQ PRNSLGFCDT NSKGLFQVLA GGTVLQLQRG DEVWIEKDPA KGRIYQGTEA DSIFSGFLIF PSA | |
Greater than 90% as determined by SDS-PAGE. |
298566 | |
10 μg | |
Not determined. | |
Full Length | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |