missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CD106 / VCAM1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) CD106 / VCAM1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8877-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_037021.1 | |
CD106 / VCAM1 Protein | |
Research Use Only | |
VCAM1 | |
Recombinant Protein | |
E. coli | |
PEYKTLAQIG DSMLLTCSTT GCESPSFSWR TQIDSPLNGK VKTEGAKSVL TMDPVSFENE HSYLCTATCN SGKLERGIQV DIYSFPKDPE IQFSGPLEVG KPVMVKCLAP DVYPIDRLEI ELFKGDRLMK KQDFVDEMAK KSLETKSLEV IFTPVIEDIE KALVCRAKLY IDQTDSIPKE RETVRELQVY TSPKNTEISV HPSTRLHEGA AVTMTCASEG LPAPEIFWSK KLDNGVLQLL SGNATLTLIA MRMEDSGIYV CEGVNLVGRD KTEVELIVQE KPFTVDISPG SQVAAQVGDS VVLTCAAVGC DSPSFSWRTQ TDSPLNGEVR DEGATSTLTL SPVGVEDEHS YLCTVTCQRR KLEKTIQVEV YSFPEDPEIE ISGPLVHGRP VTVNCTVPNV YPFDHLEIEL LKGETTLLNK FLREEIGTKS LETKSLEMTF IPTAEDTGKA LVCLAKLHSS QMESEPKQRQ STQTLYVNVA PKEPTIWVSP SPVPEEGSPV NLTCSSDGFP TPKILWSRQL KNGELQPLSQ NTTLSFMATK MEDSGIYVCE GINEAGISKK SVELIIQGSS KDIQLTVFPS KSVKEGDTVI ISCTCGSVPE IWIILKKKAK TGDMVLKSVN GSYTIRKAQL QDAGVYECES KTEVGSQLRS LTLDVKGKEN NKDYFSP | |
Greater than 90% as determined by SDS-PAGE. |
25361 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |