missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Batf3 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Batf3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5704-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_068637.1 | |
Batf3 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
60462 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Batf3 | |
Recombinant Protein | |
Yeast | |
MSQGPPAGGV LQSSVAAPGN QPQSPKDDDR KVRRREKNRV AAQRSRKKQT QKSDKLHEEH ESLEQENSVL RREIAKLKEE LRHLTEALKE HEKMCPLLLC PMNFVQLRPD PVASWSAHDA PDHPSFIWLG TLV |