missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat ACRP30 Protein
A cDNA sequence encoding the ACRP30 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10948-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
246253 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human (HEK293) | |
GITATEGPGA LVPPPKETCA GWMAGIPGYP GHNGIPGRDG RDGTPGEKGE KGDAGVLGPK GDPGDAGMTG AEGPRGFPGT PGRKGEPGEA AYMYHSAFSV GLETRVTVPN VPIRFTKIFY NQQNHYDGST GKFHCNIPGL YYFSYHITVY MKDVKVSLFK KDKAVLFTYD QYQEKNVDQA SGSMLLHLEV GDQVWLQVYG EGDNNGLYAD NVNDSTFTGF LLYHDTNAAA DYKDDDDK | |
Greater than 90.0% as determined by SDS-PAGE. |
ACRP30 Protein | |
Research Use Only | |
Adipoq | |
Rat | |
Untagged | |
Filtered (0.4 microm) and lyophilized in 20 mM Tris buffer, 50 mM NaCl, pH 7.5. |